Assay Details
| Gene Symbol: Pxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011223.3 | NP_035353.1 | 9 - 10 | 1436 | 56 | |
| NM_133915.3 | NP_598676.2 | 10 - 11 | 1538 | 56 | ||
| GenBank mRNA | AF293882.1 | - | 9 - 10 | 1361 | 56 | |
| AF293883.1 | - | 9 - 10 | 1358 | 56 | ||
| AK148060.1 | - | 9 - 10 | 1422 | 56 | ||
| AK149933.1 | - | 9 - 10 | 1401 | 56 | ||
| AK167299.1 | - | 9 - 10 | 1375 | 56 | ||
| AK171226.1 | - | 9 - 10 | 1424 | 56 | ||
| AK171436.1 | - | 9 - 10 | 1401 | 56 | ||
| AK218322.1 | - | 1 - 2 | 59 | 56 | ||
| BC003298.1 | - | 5 - 6 | 719 | 56 | ||
| BC025493.1 | - | 9 - 10 | 1394 | 56 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK089534.1 | - | Chr. - - [-] on Build | 56 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Pxn
Genomic Map