Assay Details
| Gene Symbol: Rasgrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011246.2 | NP_035376.1 | 9 - 10 | 1410 | 120 | |
| GenBank mRNA | AF106070.1 | - | 9 - 10 | 1254 | 120 | |
| AK028308.1 | - | 2 - 3 | 401 | 120 | ||
| AK047613.1 | - | 4 - 5 | 712 | 120 | ||
| AK134005.1 | - | 9 - 10 | 1423 | 120 | ||
| AK141524.1 | - | 9 - 10 | 1535 | 120 | ||
| BC057120.1 | - | 9 - 10 | 1410 | 6831137 | 120 | |
| BC057341.1 | - | 9 - 10 | 1410 | 6831137 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rasgrp1
Genomic Map