Assay Details
| Gene Symbol: Rasgrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011246.2 | NP_035376.1 | 10 - 11 | 1496 | 59 | |
| GenBank mRNA | AF106070.1 | - | 10 - 11 | 1340 | 59 | |
| AK028308.1 | - | 3 - 4 | 487 | 59 | ||
| AK047613.1 | - | 5 - 6 | 798 | 59 | ||
| AK134005.1 | - | 10 - 11 | 1509 | 59 | ||
| AK141524.1 | - | 10 - 11 | 1621 | 59 | ||
| BC057120.1 | - | 10 - 11 | 1496 | 6831137 | 59 | |
| BC057341.1 | - | 10 - 11 | 1496 | 6831137 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rasgrp1
Genomic Map