| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800017992.mm.1 |
NM_001025589.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017992.mm.1 |
NM_001025589.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017992.mm.1 |
NM_011266.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017992.mm.1 |
NM_011266.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017992.mm.1 |
TC0800002385.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800017991.mm.1 |
NM_001025589.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017991.mm.1 |
NM_001025589.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017991.mm.1 |
NM_011266.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017991.mm.1 |
NM_011266.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017991.mm.1 |
TC0800002385.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800017990.mm.1 |
NM_001025589.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017990.mm.1 |
NM_001025589.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017990.mm.1 |
NM_011266.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017990.mm.1 |
NM_011266.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017990.mm.1 |
TC0800002385.mm.1 |
MTA TC |