Assay Details
| Gene Symbol: Rfxank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011266.2 | NP_035396.1 | 4 - 5 | 648 | 56 | |
| GenBank mRNA | AF094761.1 | - | 4 - 5 | 564 | 56 | |
| AF123704.1 | - | 4 - 5 | 549 | 56 | ||
| L43164.1 | - | 4 - 5 | 549 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfxank
Genomic Map