Assay Details
| Gene Symbol: Rfxank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025589.1 | NP_001020760.1 | 6 - 7 | 847 | 91 | |
| NM_011266.2 | NP_035396.1 | 6 - 7 | 877 | 91 | ||
| GenBank mRNA | AF094761.1 | - | 6 - 7 | 793 | 91 | |
| AK052298.1 | - | 6 - 7 | 804 | 91 | ||
| AK087462.1 | - | 6 - 7 | 778 | 91 | ||
| AK162595.1 | - | 2 - 3 | 465 | 91 | ||
More Information
Set Membership: |
|

Rfxank
Genomic Map