Assay Details
| Gene Symbol: Rfxank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025589.1 | NP_001020760.1 | 7 - 8 | 914 | 98 | |
| NM_011266.2 | NP_035396.1 | 7 - 8 | 944 | 98 | ||
| GenBank mRNA | AF094761.1 | - | 7 - 8 | 860 | 98 | |
| AF123704.1 | - | 7 - 8 | 845 | 98 | ||
| AK052298.1 | - | 7 - 8 | 871 | 98 | ||
| AK087462.1 | - | 7 - 8 | 845 | 98 | ||
| AK146148.1 | - | 7 - 8 | 843 | 98 | ||
| AK162595.1 | - | 3 - 4 | 532 | 98 | ||
| BC010971.1 | - | 7 - 8 | 848 | 4190691 | 98 | |
| L43164.1 | - | 7 - 8 | 845 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfxank
Genomic Map