Assay Details
| Gene Symbol: Slc7a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253679.1 | NP_001240608.1 | 7 - 8 | 1249 | 57 | |
| NM_001253680.1 | NP_001240609.1 | 6 - 7 | 1267 | 57 | ||
| NM_011405.4 | NP_035535.2 | 8 - 9 | 1406 | 57 | ||
| GenBank mRNA | AJ012754.1 | - | 6 - 7 | 1257 | 57 | |
| AJ130943.1 | - | 7 - 8 | 1249 | 57 | ||
| AK150950.1 | - | 8 - 9 | 1406 | 57 | ||
| AK151718.1 | - | 8 - 9 | 1406 | 57 | ||
| AK152637.1 | - | 6 - 7 | 1172 | 57 | ||
| AK170851.1 | - | 6 - 7 | 1178 | 57 | ||
| BC014709.1 | - | 6 - 7 | 1255 | 4482816 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a7
Genomic Map