Assay Details
| Gene Symbol: Strn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011500.2 | NP_035630.2 | 11 - 12 | 1571 | 100 | |
| GenBank mRNA | AJ223777.1 | - | 11 - 12 | 1498 | 100 | |
| AK135684.1 | - | 6 - 7 | 764 | 100 | ||
| AK136032.1 | - | 9 - 10 | 1220 | 100 | ||
| BC090968.1 | - | 11 - 12 | 1571 | 30646517 | 100 | |
| BC150727.1 | - | 11 - 12 | 1539 | 9087638 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Strn
Genomic Map