Assay Details
| Gene Symbol: Sdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011519.2 | NP_035649.1 | 3 - 4 | 870 | 79 | |
| GenBank mRNA | AF134897.1 | - | 3 - 4 | 498 | 79 | |
| AK132236.1 | - | 3 - 4 | 912 | 79 | ||
| AK132491.1 | - | 3 - 4 | 853 | 79 | ||
| AK143529.1 | - | 3 - 4 | 882 | 79 | ||
| AK162092.1 | - | 3 - 4 | 855 | 79 | ||
| AK168091.1 | - | 3 - 4 | 759 | 79 | ||
| BC010560.1 | - | 3 - 4 | 892 | 3501359 | 79 | |
| X15487.1 | - | 3 - 4 | 869 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sdc1
Genomic Map