Assay Details
| Gene Symbol: Tfeb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161722.1 | NP_001155194.1 | 2 - 3 | 512 | 100 | |
| NM_001161723.1 | NP_001155195.1 | 2 - 3 | 506 | 100 | ||
| NM_011549.3 | NP_035679.3 | 1 - 2 | 453 | 100 | ||
| GenBank mRNA | AF079095.1 | - | 1 - 2 | 434 | 100 | |
| AK078755.1 | - | 2 - 3 | 444 | 100 | ||
| AK146016.1 | - | 1 - 2 | 478 | 100 | ||
| BC064789.1 | - | 1 - 2 | 453 | 30429545 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tfeb
Genomic Map