Assay Details
| Gene Symbol: Tead2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285498.1 | NP_001272427.1 | 8 - 9 | 865 | 79 | |
| NM_001285500.1 | NP_001272429.1 | 8 - 9 | 718 | 79 | ||
| NM_011565.2 | NP_035695.1 | 8 - 9 | 823 | 79 | ||
| GenBank mRNA | AK132009.1 | - | 7 - 8 | 758 | 79 | |
| AK137064.1 | - | 7 - 8 | 2106 | 79 | ||
| AK143155.1 | - | 7 - 8 | 2039 | 79 | ||
| AK144857.1 | - | 8 - 9 | 785 | 79 | ||
| AK146595.1 | - | 8 - 9 | 785 | 79 | ||
| BC020330.1 | - | 2 - 3 | 220 | 79 | ||
| BC050217.1 | - | 7 - 8 | 643 | 30021393 | 79 | |
| D50563.1 | - | 8 - 9 | 823 | 79 | ||
| X94442.1 | - | 7 - 8 | 737 | 79 | ||
| Y10026.1 | - | 8 - 9 | 787 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tead2
Genomic Map