Assay Details
| Gene Symbol: Tead3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098226.3 | NP_001091696.2 | 9 - 10 | 823 | 68 | |
| NM_001204156.1 | NP_001191085.1 | 9 - 10 | 1146 | 68 | ||
| NM_011566.4 | NP_035696.3 | 9 - 10 | 987 | 68 | ||
| GenBank mRNA | AF002670.1 | - | 9 - 10 | 942 | 68 | |
| AK087649.1 | - | 9 - 10 | 825 | 68 | ||
| AK143551.1 | - | 8 - 9 | 1859 | 68 | ||
| BC021364.1 | - | 9 - 10 | 867 | 68 | ||
| BC047929.1 | - | 9 - 10 | 1146 | 68 | ||
| D87963.1 | - | 8 - 9 | 657 | 68 | ||
| D87964.1 | - | 9 - 10 | 968 | 68 | ||
| Y10027.1 | - | 8 - 9 | 749 | 68 | ||
| Y16611.1 | - | 8 - 9 | 740 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tead3
Genomic Map