Assay Details
| Gene Symbol: Top3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011624.2 | NP_035754.1 | 4 - 5 | 589 | 72 | |
| GenBank mRNA | AB013603.1 | - | 3 - 4 | 402 | 72 | |
| AB045324.1 | - | 4 - 5 | 663 | 72 | ||
| AK041159.1 | - | 4 - 5 | 499 | 72 | ||
| AK144709.1 | - | 5 - 6 | 575 | 72 | ||
| AK146475.1 | - | 4 - 5 | 611 | 72 | ||
| AK180059.1 | - | 1 - 2 | 69 | 72 | ||
| BC006614.1 | - | 4 - 5 | 492 | 72 | ||
| BC031723.1 | - | 4 - 5 | 589 | 5003002 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Top3b
Genomic Map