Assay Details
| Gene Symbol: Top3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011624.2 | NP_035754.1 | 14 - 15 | 1932 | 70 | |
| GenBank mRNA | AB013603.1 | - | 13 - 14 | 1745 | 70 | |
| AK144709.1 | - | 15 - 16 | 1918 | 70 | ||
| AK146475.1 | - | 14 - 15 | 1954 | 70 | ||
| AK163796.1 | - | 7 - 8 | 942 | 70 | ||
| BC031723.1 | - | 14 - 15 | 1932 | 5003002 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Top3b
Genomic Map