Assay Details
| Gene Symbol: Top3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011624.2 | NP_035754.1 | 15 - 16 | 2082 | 90 | |
| GenBank mRNA | AB013603.1 | - | 14 - 15 | 1895 | 90 | |
| AK144709.1 | - | 16 - 17 | 2068 | 90 | ||
| AK146475.1 | - | 15 - 16 | 2104 | 90 | ||
| AK163716.1 | - | 2 - 3 | 944 | 90 | ||
| BC031723.1 | - | 15 - 16 | 2082 | 5003002 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Top3b
Genomic Map