Assay Details
| Gene Symbol: Top3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011624.2 | NP_035754.1 | 16 - 17 | 2188 | 83 | |
| GenBank mRNA | AB013603.1 | - | 15 - 16 | 2001 | 83 | |
| AB045324.1 | - | 15 - 16 | 2738 | 83 | ||
| AK144709.1 | - | 17 - 18 | 2174 | 83 | ||
| AK146475.1 | - | 16 - 17 | 2210 | 83 | ||
| AK163716.1 | - | 3 - 4 | 1050 | 83 | ||
| AK163796.1 | - | 9 - 10 | 1197 | 83 | ||
| BC031723.1 | - | 16 - 17 | 2188 | 5003002 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Top3b
Genomic Map