Assay Details
| Gene Symbol: Vars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011690.3 | NP_035820.3 | 13 - 14 | 2027 | 59 | |
| GenBank mRNA | AF087680.1 | - | 13 - 14 | 1790 | 59 | |
| AK019157.1 | - | 5 - 6 | 424 | 59 | ||
| AK154823.1 | - | 13 - 14 | 1842 | 59 | ||
| AK157814.1 | - | 13 - 14 | 1885 | 59 | ||
| AK165326.1 | - | 12 - 13 | 1470 | 59 | ||
| AK167842.1 | - | 5 - 6 | 565 | 59 | ||
| BC053703.1 | - | 13 - 14 | 1810 | 30055190 | 59 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Vars
Genomic Map