Assay Details
Gene Symbol: Trpv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011706.2 | NP_035836.2 | 8 - 9 | 1616 | 87 | |
GenBank mRNA | AB021665.1 | - | 8 - 9 | 1698 | 87 | |
AK008689.1 | - | 6 - 7 | 1141 | 87 | ||
AK089004.1 | - | 9 - 10 | 1631 | 87 | ||
BC005415.1 | - | 7 - 8 | 1563 | 3487527 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
