Assay Details
| Gene Symbol: Pde10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290707.1 | NP_001277636.1 | 5 - 6 | 722 | 70 | |
| NM_011866.2 | NP_035996.2 | 4 - 5 | 676 | 70 | ||
| GenBank mRNA | AF110507.1 | - | 4 - 5 | 547 | 70 | |
| AK039249.1 | - | 5 - 6 | 722 | 70 | ||
| AK162221.1 | - | 4 - 5 | 331 | 70 | ||
| AK162804.1 | - | 2 - 3 | 217 | 70 | ||
| AY360383.1 | - | 4 - 5 | 676 | 70 | ||
| BC113201.1 | - | 4 - 5 | 542 | 40048238 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde10a
Genomic Map