Assay Details
| Gene Symbol: Ptpn21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011877.2 | NP_036007.2 | 1 - 2 | 178 | 86 | |
| GenBank mRNA | AK013777.1 | - | 1 - 2 | 180 | 86 | |
| AK030456.1 | - | 1 - 2 | 179 | 86 | ||
| D37801.1 | - | 1 - 2 | 127 | 86 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK030456.1,D37801.1,NM_011877.2 transcripts.
Set Membership: |
|

Ptpn21
Genomic Map