Assay Details
| Gene Symbol: Sh3bp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011894.2 | NP_036024.2 | 4 - 5 | 577 | 74 | |
| GenBank mRNA | AB016835.1 | - | 4 - 5 | 405 | 74 | |
| AK043375.1 | - | 622 | 74 | |||
| AK161427.1 | - | 3 - 4 | 559 | 74 | ||
| AK165033.1 | - | 4 - 5 | 611 | 74 | ||
| BC018237.1 | - | 4 - 5 | 609 | 3482670 | 74 | |
| BC053741.1 | - | 4 - 5 | 577 | 6336511 | 74 | |
More Information
Set Membership: |
|

Sh3bp5
Genomic Map