Assay Details
| Gene Symbol: Tlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294330.1 | NP_001281259.1 | 5 - 6 | 790 | 90 | |
| NM_001294331.1 | NP_001281260.1 | 6 - 7 | 899 | 90 | ||
| GenBank mRNA | AF045252.1 | - | 6 - 7 | 905 | 90 | |
| BC066198.1 | - | 5 - 6 | 335 | 90 | ||
More Information
Set Membership: |
|

Tlk2
Genomic Map