Assay Details
| Gene Symbol: Tlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112705.2 | NP_001106176.1 | 10 - 11 | 1360 | 80 | |
| NM_001294329.1 | NP_001281258.1 | 8 - 9 | 1220 | 80 | ||
| NM_001294330.1 | NP_001281259.1 | 8 - 9 | 1168 | 80 | ||
| NM_001294331.1 | NP_001281260.1 | 10 - 11 | 1373 | 80 | ||
| NM_001294334.1 | NP_001281263.1 | 10 - 11 | 1360 | 80 | ||
| GenBank mRNA | AF045252.1 | - | 10 - 11 | 1379 | 80 | |
| AF045253.1 | - | 9 - 10 | 807 | 80 | ||
| AF045254.1 | - | 5 - 6 | 521 | 80 | ||
| AK014829.1 | - | 10 - 11 | 998 | 80 | ||
| AK165667.1 | - | 10 - 11 | 1363 | 80 | ||
| AX775743.1 | - | 4 - 5 | 385 | 80 | ||
| BC066198.1 | - | 8 - 9 | 713 | 80 | ||
| CD554252.1 | - | 4 - 5 | 484 | 80 | ||
| U56909.1 | - | 1 - 2 | 58 | 80 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the U56909.1 transcripts.
Set Membership: |
|

Tlk2
Genomic Map