Assay Details
| Gene Symbol: Tlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112705.2 | NP_001106176.1 | 11 - 12 | 1486 | 87 | |
| NM_001294329.1 | NP_001281258.1 | 9 - 10 | 1346 | 87 | ||
| NM_001294330.1 | NP_001281259.1 | 9 - 10 | 1294 | 87 | ||
| NM_001294331.1 | NP_001281260.1 | 11 - 12 | 1499 | 87 | ||
| NM_001294334.1 | NP_001281263.1 | 11 - 12 | 1486 | 87 | ||
| GenBank mRNA | AF045252.1 | - | 11 - 12 | 1505 | 87 | |
| AF045253.1 | - | 10 - 11 | 933 | 87 | ||
| AF045254.1 | - | 6 - 7 | 647 | 87 | ||
| AK014829.1 | - | 11 - 12 | 1124 | 87 | ||
| AK089925.1 | - | 9 - 10 | 874 | 87 | ||
| AK165667.1 | - | 11 - 12 | 1489 | 87 | ||
| AX775743.1 | - | 5 - 6 | 511 | 87 | ||
| BC066198.1 | - | 9 - 10 | 839 | 87 | ||
| U56909.1 | - | 2 - 3 | 184 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tlk2
Genomic Map