Assay Details
| Gene Symbol: Usp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011909.2 | NP_036039.2 | 9 - 10 | 1279 | 65 | |
| GenBank mRNA | AF069502.1 | - | 9 - 10 | 1240 | 65 | |
| AK149772.1 | - | 9 - 10 | 1280 | 65 | ||
| AK151011.1 | - | 9 - 10 | 1280 | 65 | ||
| AK151260.1 | - | 9 - 10 | 1280 | 65 | ||
| AK151722.1 | - | 9 - 10 | 1280 | 65 | ||
| AK152034.1 | - | 9 - 10 | 1245 | 65 | ||
| AK152574.1 | - | 9 - 10 | 1245 | 65 | ||
| AK153312.1 | - | 9 - 10 | 1249 | 65 | ||
| BC138562.1 | - | 9 - 10 | 1161 | 8861582 | 65 | |
| BC138577.1 | - | 9 - 10 | 1161 | 8861597 | 65 | |
| CT010228.1 | - | 8 - 9 | 1015 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp18
Genomic Map