Assay Details
| Gene Symbol: Parg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011960.2 | NP_036090.2 | 6 - 7 | 1915 | 102 | |
| GenBank mRNA | AF079557.1 | - | 6 - 7 | 1664 | 102 | |
| AK036656.1 | - | 6 - 7 | 1938 | 102 | ||
| AK088708.1 | - | 3 - 4 | 270 | 102 | ||
| AK160742.1 | - | 5 - 6 | 1389 | 102 | ||
| AK163388.1 | - | 6 - 7 | 1942 | 102 | ||
| AK164013.1 | - | 6 - 7 | 1902 | 102 | ||
| BC050892.1 | - | 6 - 7 | 1914 | 102 | ||
| BC053037.1 | - | 6 - 7 | 1898 | 102 | ||
| BC059827.1 | - | 6 - 7 | 1924 | 102 | ||
| EF382673.1 | - | 4 - 5 | 624 | 102 | ||
| JQ890227.1 | - | 3 - 4 | 421 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Parg
Genomic Map