Assay Details
| Gene Symbol: Parg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011960.2 | NP_036090.2 | 12 - 13 | 2465 | 82 | |
| GenBank mRNA | AF079557.1 | - | 12 - 13 | 2214 | 82 | |
| AK034542.1 | - | 5 - 6 | 397 | 82 | ||
| AK036656.1 | - | 12 - 13 | 2488 | 82 | ||
| AK088708.1 | - | 9 - 10 | 820 | 82 | ||
| AK164013.1 | - | 12 - 13 | 2452 | 82 | ||
| BC050892.1 | - | 12 - 13 | 2464 | 82 | ||
| BC053037.1 | - | 12 - 13 | 2449 | 82 | ||
| BC059827.1 | - | 12 - 13 | 2474 | 82 | ||
| EF382673.1 | - | 10 - 11 | 1174 | 82 | ||
| JQ890227.1 | - | 9 - 10 | 971 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Parg
Genomic Map