Assay Details
| Gene Symbol: Pla2g10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291009.1 | NP_001277938.1 | 1 - 2 | 627 | 139 | |
| NM_011987.3 | NP_036117.1 | 1 - 2 | 627 | 139 | ||
| NR_110990.1 | 1 - 2 | 627 | 139 | |||
| GenBank mRNA | AF166097.2 | - | 1 - 2 | 229 | 139 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF166097.2,NM_001291009.1,NM_011987.3 transcripts.
Set Membership: |
|

Pla2g10
Genomic Map