Assay Details
| Gene Symbol: Omd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012050.2 | NP_036180.1 | 3 - 4 | 1204 | 106 | |
| GenBank mRNA | AB007848.1 | - | 3 - 4 | 1209 | 106 | |
| AK050297.1 | - | 1 - 2 | 307 | 106 | ||
| BC095997.1 | - | 2 - 3 | 1010 | 30739852 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342926 | NM_012050.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342926 | AB007848.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342926 | NM_012050.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342926 | AB007848.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109500 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342926 | NM_012050.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342926 | AB007848.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342926 | NM_012050.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342926 | AB007848.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342926 | NM_012050.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342926 | AB007848.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342926 | NM_012050.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342926 | AB007848.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29026100 | - | - |
| Whole Mouse Genome G4122A | A_51_P342926 | NM_012050.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342926 | AB007848.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342926 | NM_012050.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342926 | AB007848.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107162 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11116537 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11116529 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11116532 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11116535 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11116549 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11116544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29026123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10467150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10526452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11007344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10554394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11116546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29026120 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10405059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405060 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405061 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418745_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10405058 | - | - |
| Mouse Genome 430 2.0 Array | 1418745_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418745_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27047_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27047_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300003512.mm.1 | NM_012050.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003512.mm.1 | NM_012050.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003512.mm.1 | TC1300000568.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300003510.mm.1 | NM_012050.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003510.mm.1 | NM_012050.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003510.mm.1 | TC1300000568.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300003511.mm.1 | NM_012050.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003511.mm.1 | NM_012050.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003511.mm.1 | TC1300000568.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300003513.mm.1 | NM_012050.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003513.mm.1 | NM_012050.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003513.mm.1 | TC1300000568.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418745_at | - | - |
| Mouse Expression Set 430 | 1418745_PM_at | - | - |
| Murine Genome U74v2 Set | 93369_PM_at | - | - |
| Murine Genome U74v2 Set | 93369_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418745_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418745_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418745_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418745_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 558014 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6110044 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6110044 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110044 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1238597 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238597 | - | - |
| mouse-6 v1.1 (Illumina) | scl44789.4.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110044 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44789.4.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110044 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1238597 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238597 | - | - |

Omd
Genomic Map