Assay Details
Gene Symbol: Olfr64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013617.3 | NP_038645.2 | 1 | 855 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994867 | NM_013617.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994867 | NM_013617.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P7221 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385600 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385597 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385587 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385598 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385590 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385582 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385570 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385575 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385585 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385574 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385577 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385604 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385602 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385594 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385580 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385603 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385586 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385567 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385605 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385573 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385578 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385595 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385592 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385583 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385589 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25385568 | - | - |
Whole Mouse Genome G4122A | A_52_P7221 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136176 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06307891 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05957454 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06022687 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06022672 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05957449 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05935439 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05935437 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05935447 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05935443 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06022670 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05935450 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05957447 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05935442 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06022666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25385573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06414917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385583 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385590 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385592 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05935448 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385585 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385574 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385559 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25385575 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10566285 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566284 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1422386_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10566283 | - | - |
Mouse Genome 430 2.0 Array | 1422374_s_at | - | - |
Mouse Genome 430 2.0 Array | 1422374_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1422386_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1422386_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18366_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18366_at | - | - |
Mouse Expression Set 430 | 1422386_at | - | - |
Mouse Expression Set 430 | 1422386_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422386_at | - | - |
Mouse Genome 430A 2.0 Array | 1422386_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422386_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422386_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 510670 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000510670 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1256949 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1256949 | - | - |
mouse-6 v1.1 (Illumina) | scl30920.1.1_82-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 510670 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl30920.1.1_82-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510670 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2965952 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256949 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2965952 | - | - |
