Assay Details
| Gene Symbol: Pam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013626.3 | NP_038654.2 | 14 - 15 | 1622 | 67 | |
| GenBank mRNA | U79523.1 | - | 14 - 15 | 1507 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pam
Genomic Map