Assay Details
| Gene Symbol: Supt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013676.1 | NP_038704.1 | 13 - 14 | 1066 | 65 | |
| GenBank mRNA | AK145117.1 | - | 13 - 14 | 1106 | 65 | |
| AK146055.1 | - | 13 - 14 | 1094 | 65 | ||
| AK146453.1 | - | 13 - 14 | 1094 | 65 | ||
| AK146583.1 | - | 12 - 13 | 1313 | 65 | ||
| AK146650.1 | - | 13 - 14 | 1107 | 65 | ||
| AK159786.1 | - | 14 - 15 | 1191 | 65 | ||
| BC007132.1 | - | 13 - 14 | 1066 | 3496691 | 65 | |
| BC057449.1 | - | 13 - 14 | 1081 | 65 | ||
| BC058598.1 | - | 12 - 13 | 1041 | 6306109 | 65 | |
| BC059849.1 | - | 5 - 6 | 649 | 5696323 | 65 | |
| U88539.1 | - | 13 - 14 | 1093 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt5
Genomic Map