Assay Details
| Gene Symbol: Syn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110780.1 | NP_001104250.1 | 1 - 2 | 509 | 68 | |
| NM_013680.4 | NP_038708.3 | 1 - 2 | 509 | 68 | ||
| GenBank mRNA | BB646141.1 | - | 1 - 2 | 510 | 68 | |
| BC022954.1 | - | 1 - 2 | 487 | 5368098 | 68 | |
More Information
Set Membership: |
|

Syn1
Genomic Map