Assay Details
| Gene Symbol: Syn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110780.1 | NP_001104250.1 | 5 - 6 | 906 | 104 | |
| NM_013680.4 | NP_038708.3 | 5 - 6 | 906 | 104 | ||
| GenBank mRNA | BC022954.1 | - | 5 - 6 | 884 | 5368098 | 104 |
More Information
Updated Mapping Information
Set Membership: |
|

Syn1
Genomic Map