Assay Details
| Gene Symbol: Klf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289471.1 | NP_001276400.1 | 2 - 3 | 381 | 77 | |
| NM_013692.3 | NP_038720.1 | 2 - 3 | 381 | 77 | ||
| GenBank mRNA | AF049879.1 | - | 2 - 3 | 377 | 77 | |
| AF064088.1 | - | 2 - 3 | 373 | 77 | ||
| AK004906.1 | - | 2 - 3 | 380 | 77 | ||
| AK043433.1 | - | 2 - 3 | 377 | 77 | ||
| AK079679.1 | - | 2 - 3 | 377 | 77 | ||
| AK145676.1 | - | 2 - 3 | 253 | 77 | ||
| AK154719.1 | - | 2 - 3 | 377 | 77 | ||
| BC003316.1 | - | 2 - 3 | 321 | 3155247 | 77 | |
| Z36270.1 | - | 2 - 3 | 379 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klf10
Genomic Map