Assay Details
| Gene Symbol: Podxl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013723.3 | NP_038751.2 | 3 - 4 | 1142 | 57 | |
| GenBank mRNA | AB028048.1 | - | 3 - 4 | 1023 | 57 | |
| AF290209.1 | - | 3 - 4 | 1041 | 57 | ||
| BC052442.1 | - | 3 - 4 | 1116 | 6406610 | 57 | |
| BC054530.1 | - | 3 - 4 | 1116 | 6406610 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Podxl
Genomic Map