Assay Details
| Gene Symbol: Podxl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013723.3 | NP_038751.2 | 5 - 6 | 1374 | 61 | |
| GenBank mRNA | AB028048.1 | - | 5 - 6 | 1255 | 61 | |
| AF290209.1 | - | 5 - 6 | 1273 | 61 | ||
| AK134723.1 | - | 1 - 2 | 110 | 61 | ||
| BC052442.1 | - | 5 - 6 | 1348 | 6406610 | 61 | |
| BC054530.1 | - | 5 - 6 | 1348 | 6406610 | 61 | |
More Information
Set Membership: |
|

Podxl
Genomic Map