Assay Details
| Gene Symbol: Sh2d3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252547.1 | NP_001239476.1 | 5 - 6 | 873 | 78 | |
| NM_013781.3 | NP_038809.1 | 6 - 7 | 1359 | 78 | ||
| NR_045535.1 | 7 - 8 | 1817 | 78 | |||
| GenBank mRNA | AB043953.1 | - | 6 - 7 | 1342 | 78 | |
| AF168364.1 | - | 6 - 7 | 1300 | 78 | ||
| AK042709.1 | - | 5 - 6 | 857 | 78 | ||
| AK085885.1 | - | 857 | 78 | |||
| AK155165.1 | - | 6 - 7 | 1295 | 78 | ||
| AK172072.1 | - | 7 - 8 | 1738 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh2d3c
Genomic Map