Assay Details
| Gene Symbol: Trove2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013835.2 | NP_038863.1 | 3 - 4 | 957 | 79 | |
| GenBank mRNA | AK076167.1 | - | 3 - 4 | 934 | 79 | |
| AK167379.1 | - | 3 - 4 | 898 | 79 | ||
| AK167555.1 | - | 3 - 4 | 898 | 79 | ||
| AK167645.1 | - | 3 - 4 | 898 | 79 | ||
| BC040368.1 | - | 2 - 3 | 828 | 79 | ||
| BC051974.1 | - | 2 - 3 | 834 | 5697399 | 79 | |
| BC052380.1 | - | 2 - 3 | 834 | 5697399 | 79 | |
| L81154.1 | - | 2 - 3 | 805 | 79 | ||
| U66843.1 | - | 3 - 4 | 884 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trove2
Genomic Map