Assay Details
| Gene Symbol: Dpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013874.2 | NP_038902.1 | 9 - 10 | 982 | 133 | |
| GenBank mRNA | AK142419.1 | - | 8 - 9 | 1040 | 133 | |
| BC052348.1 | - | 9 - 10 | 1011 | 6587002 | 133 | |
| U48238.1 | - | 9 - 10 | 981 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpf1
Genomic Map