Assay Details
| Gene Symbol: Pde7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013875.5 | NP_038903.3 | 10 - 11 | 1319 | 66 | |
| GenBank mRNA | AF190639.1 | - | 10 - 11 | 1100 | 66 | |
| AJ251859.1 | - | 10 - 11 | 1371 | 66 | ||
| AK035385.1 | - | 10 - 11 | 1331 | 66 | ||
| AK049910.1 | - | 10 - 11 | 1375 | 66 | ||
| BC130267.1 | - | 10 - 11 | 1200 | 40143060 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde7b
Genomic Map