Assay Details
| Gene Symbol: Rnf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013876.3 | NP_038904.1 | 1 - 2 | 394 | 70 | |
| GenBank mRNA | AB024427.1 | - | 2 - 3 | 259 | 70 | |
| AF220206.1 | - | 1 - 2 | 148 | 70 | ||
| AK014254.1 | - | 1 - 2 | 548 | 70 | ||
| BC010299.1 | - | 1 - 2 | 394 | 3155804 | 70 | |
| BC028255.1 | - | 1 - 2 | 575 | 4972336 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf11
Genomic Map