Assay Details
| Gene Symbol: Scmh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159630.1 | NP_001153102.1 | 9 - 10 | 1173 | 97 | |
| NM_013883.2 | NP_038911.1 | 9 - 10 | 1173 | 97 | ||
| GenBank mRNA | AB030906.1 | - | 9 - 10 | 1072 | 97 | |
| AB221680.1 | - | 5 - 6 | 714 | 97 | ||
| AB221681.1 | - | 5 - 6 | 714 | 97 | ||
| BC034667.1 | - | 7 - 8 | 883 | 5356322 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scmh1
Genomic Map