| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600010266.mm.1 |
NM_015773.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010266.mm.1 |
NM_015773.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010266.mm.1 |
TC1600001267.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600010262.mm.1 |
NM_015773.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010262.mm.1 |
NM_015773.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010262.mm.1 |
TC1600001267.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600010270.mm.1 |
NM_015773.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010270.mm.1 |
NM_015773.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010270.mm.1 |
TC1600001267.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600010269.mm.1 |
NM_015773.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010269.mm.1 |
NM_015773.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010269.mm.1 |
TC1600001267.mm.1 |
MTA TC |