Assay Details
| Gene Symbol: Spag6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015773.2 | NP_056588.1 | 3 - 4 | 426 | 88 | |
| GenBank mRNA | AF173866.1 | - | 3 - 4 | 363 | 88 | |
| AK005732.1 | - | 3 - 4 | 374 | 88 | ||
| AK054199.1 | - | 2 - 3 | 462 | 88 | ||
| BC090660.1 | - | 3 - 4 | 369 | 30673350 | 88 | |
| BC100455.1 | - | 3 - 4 | 376 | 30919921 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag6
Genomic Map