Assay Details
| Gene Symbol: Nap1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146707.1 | NP_001140179.1 | 9 - 10 | 1597 | 104 | |
| NM_015781.4 | NP_056596.1 | 10 - 11 | 1094 | 104 | ||
| GenBank mRNA | AK004633.1 | - | 10 - 11 | 1064 | 104 | |
| AK007322.1 | - | 10 - 11 | 1059 | 104 | ||
| AK012250.1 | - | 6 - 7 | 605 | 104 | ||
| AK032896.1 | - | 10 - 11 | 1067 | 104 | ||
| AK050375.1 | - | 9 - 10 | 1599 | 104 | ||
| AK136161.1 | - | 10 - 11 | 1060 | 104 | ||
| AK145766.1 | - | 10 - 11 | 1058 | 104 | ||
| AK152011.1 | - | 10 - 11 | 988 | 104 | ||
| AK153531.1 | - | 10 - 11 | 988 | 104 | ||
| AK169301.1 | - | 10 - 11 | 1006 | 104 | ||
| AK195357.1 | - | 2 - 3 | 210 | 105 | ||
| AK216281.1 | - | 2 - 3 | 209 | 104 | ||
| BC076591.1 | - | 9 - 10 | 1145 | 30618066 | 104 | |
| D12618.1 | - | 10 - 11 | 1050 | 104 | ||
| X61449.1 | - | 7 - 8 | 767 | 104 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nap1l1
Genomic Map