Assay Details
| Gene Symbol: Scarb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016741.2 | NP_058021.1 | 11 - 12 | 1618 | 82 | |
| GenBank mRNA | AK028191.1 | - | 11 - 12 | 1599 | 82 | |
| AK033114.1 | - | 11 - 12 | 1543 | 82 | ||
| AK154933.1 | - | 11 - 12 | 1610 | 82 | ||
| AK158122.1 | - | 11 - 12 | 1543 | 82 | ||
| AK169906.1 | - | 11 - 12 | 1594 | 82 | ||
| AK204897.1 | - | 1 - 2 | 121 | 82 | ||
| BC004656.1 | - | 11 - 12 | 1598 | 3498018 | 82 | |
| CT010222.1 | - | 11 - 12 | 1406 | 82 | ||
| U37799.1 | - | 11 - 12 | 1456 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scarb1
Genomic Map