Assay Details
| Gene Symbol: Prdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016764.4 | NP_058044.1 | 1 - 2 | 338 | 72 | |
| GenBank mRNA | AK005031.1 | - | 1 - 2 | 317 | 72 | |
| AK015920.1 | - | 1 - 2 | 314 | 72 | ||
| AK146402.1 | - | 1 - 2 | 332 | 72 | ||
| AK152255.1 | - | 1 - 2 | 320 | 72 | ||
| BC003349.1 | - | 1 - 2 | 325 | 3481827 | 72 | |
| BC019578.1 | - | 1 - 2 | 353 | 2654429 | 72 | |
| U96746.1 | - | 1 - 2 | 336 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prdx4
Genomic Map