Assay Details
| Gene Symbol: Tnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110147.1 | NP_001103617.1 | 4 - 5 | 943 | 110 | |
| NM_016788.3 | NP_058068.2 | 4 - 5 | 943 | 110 | ||
| GenBank mRNA | AF037260.1 | - | 4 - 5 | 726 | 110 | |
| AK087965.1 | - | 1 - 2 | 100 | 110 | ||
| BC052421.1 | - | 4 - 5 | 943 | 6400209 | 110 | |
| DQ666696.1 | - | 4 - 5 | 754 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnk2
Genomic Map